Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Aminobenzhydrazide, MP Biomedicals

Catalog No. ICN220229 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN220229 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN220229 Supplier MP Biomedicals, Inc Supplier No. 0522022901

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5351-17-7
Molecular Formula C7H9N3O
Molecular Weight (g/mol) 151.169
InChI Key WPBZMCGPFHZRHJ-UHFFFAOYSA-N
Synonym 4-aminobenzhydrazide, amben hydrazide, p-aminobenzhydrazide, 4-aminobenzoic acid hydrazide, aminostimil, 4-aminobenzoylhydrazine, p-aminobenzoic acid hydrazide, p-aminobenzoyl hydrazide, benzoic acid, 4-amino-, hydrazide, p-aminobenzoic hydrazide
PubChem CID 21450
IUPAC Name 4-aminobenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)N

Specifications

Quantity 10 g
Formula Weight 151
Chemical Name or Material p-Aminobenzhydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.