accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 53531-69-4

IUPAC Name: (4-bromophenyl)methanesulfonyl chloride
Synonyms: (4-bromophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 269.54
Molecular Formula: C7H6BrClO2S
InChi Key: ZWVWFWGJZPHCHF-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results