Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Bromophenyl)methanesulfonyl chloride, 97%, Thermo Scientific™

Catalog No. MO00946DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00946DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO00946DA Supplier Thermo Scientific Chemicals Supplier No. MO00946DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53531-69-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.537
MDL Number MFCD04973454
InChI Key ZWVWFWGJZPHCHF-UHFFFAOYSA-N
Synonym 4-bromobenzylsulfonyl chloride, 4-bromophenyl methanesulfonyl chloride, 4-bromobenzylsulphonyl chloride, benzenemethanesulfonyl chloride, 4-bromo, 4-bromophenyl methanesulphonyl chloride, 4-bromophenyl methyl chlorosulfone, 4-bromo-phenyl-methanesulfonyl chloride, 4-bromo-benzenemethanesulfonyl chloride, pubchem2155, acmc-209l8j
PubChem CID 598764
IUPAC Name (4-bromophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 269.55
Percent Purity 95%
Chemical Name or Material (4-Bromophenyl)methanesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.