Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzylsulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog No. AC451490050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451490050 5 g
AC451490250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451490050 Supplier Thermo Scientific Chemicals Supplier No. 451490050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 53531-69-4 | C7H6BrClO2S | 269.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53531-69-4
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.55
InChI Key ZWVWFWGJZPHCHF-UHFFFAOYSA-N
Synonym 4-bromobenzylsulfonyl chloride, 4-bromophenyl methanesulfonyl chloride, 4-bromobenzylsulphonyl chloride, benzenemethanesulfonyl chloride, 4-bromo, 4-bromophenyl methanesulphonyl chloride, 4-bromophenyl methyl chlorosulfone, 4-bromo-phenyl-methanesulfonyl chloride, 4-bromo-benzenemethanesulfonyl chloride, pubchem2155, acmc-209l8j
PubChem CID 598764
IUPAC Name (4-bromophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC=C1CS(=O)(=O)Cl)Br

Specifications

Melting Point 116.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 269.55
Percent Purity 95%
Chemical Name or Material 4-Bromobenzylsulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.