accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNH

CAS RN 53617-36-0

IUPAC Name: 1-methyl-4-(piperidin-4-yl)piperazine
Synonyms: 1-methyl-4-(piperidin-4-yl)piperazine
Molecular Weight (g/mol): 183.3
Molecular Formula: C10H21N3
InChi Key: MRYYJGQKVGZGSB-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results