accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3HOOOCH3OHOHO

CAS RN 537-09-7

IUPAC Name: 2-hydroxy-4-(2-hydroxy-4-methoxy-6-methylbenzoyloxy)-6-methylbenzoic acid
Synonyms: evernic acid
Molecular Weight (g/mol): 332.31
Molecular Formula: C17H16O7
InChi Key: GODLCSLPZIBRMG-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C(=O)OC2=CC(C)=C(C(O)=O)C(O)=C2)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results