accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 5379-77-1

IUPAC Name: 5,11-diphenyl-4,10-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 5,11-diphenyl-4,10-dioxatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 310.35
Molecular Formula: C22H14O2
InChi Key: QDHGBWBEBOMJCI-UHFFFAOYSA-N
SMILES: O1C2=CC3=C(OC(=C3)C3=CC=CC=C3)C=C2C=C1C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results