CAS RN 538-09-0
IUPAC Name:
8-azabicyclo[3.2.1]octan-3-ol
Synonyms:
8-azabicyclo[3.2.1]octan-3-ol
Molecular Weight (g/mol):
127.19
Molecular Formula:
C7H13NO
InChi Key:
YYMCYJLIYNNOMK-UHFFFAOYNA-N
SMILES:
OC1CC2CCC(C1)N2
1
–
1
of
1
results
1
Nortropine 98.0+%, TCI America™
CAS: 538-09-0 Molecular Formula: C7H13NO Molecular Weight (g/mol): 127.19 MDL Number: MFCD00047140,MFCD01711246,MFCD21642368 InChI Key: YYMCYJLIYNNOMK-UHFFFAOYNA-N Synonym: 8-Azabicyclo[3.2.1]octan-3-ol PubChem CID: 73480 IUPAC Name: 8-azabicyclo[3.2.1]octan-3-ol SMILES: OC1CC2CCC(C1)N2