CAS RN 538-75-0
N,N'-Dicyclohexylcarbodiimide, 99%
CAS: 538-75-0 Molecular Formula: C13H22N2 Molecular Weight (g/mol): 206.333 MDL Number: MFCD00011659 InChI Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N Synonym: dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl PubChem CID: 10868 ChEBI: CHEBI:53090 IUPAC Name: N,N'-dicyclohexylmethanediimine SMILES: C1CCC(CC1)N=C=NC2CCCCC2
N,N'-Dicyclohexylcarbodiimide 98.0+%, TCI America™
CAS: 538-75-0 Molecular Formula: C13H22N2 Molecular Weight (g/mol): 206.333 MDL Number: MFCD00011659 InChI Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N Synonym: dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl PubChem CID: 10868 ChEBI: CHEBI:53090 IUPAC Name: N,N'-dicyclohexylmethanediimine SMILES: C1CCC(CC1)N=C=NC2CCCCC2
N,N'-Dicyclohexylcarbodiimide, 99%
CAS: 538-75-0 Molecular Formula: C13H22N2 Molecular Weight (g/mol): 206.33 MDL Number: MFCD00011659 InChI Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N Synonym: dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl PubChem CID: 10868 ChEBI: CHEBI:53090 IUPAC Name: N,N'-dicyclohexylmethanediimine SMILES: C1CCC(CC1)N=C=NC2CCCCC2
N,N'-Dicyclohexylcarbodiimide, 1M solution in dichloromethane, AcroSeal™
CAS: 538-75-0 | C13H22N2 | 206.33 g/mol
N,N'-Dicyclohexylcarbodiimide (25% in Pyridine, ca. 1.2mol/L), TCI America™
CAS: 538-75-0 Molecular Formula: C13H22N2 Molecular Weight (g/mol): 206.333 MDL Number: MFCD00011659 InChI Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N Synonym: dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl PubChem CID: 10868 ChEBI: CHEBI:53090 IUPAC Name: N,N'-dicyclohexylmethanediimine SMILES: C1CCC(CC1)N=C=NC2CCCCC2
Sigma Aldrich Dcc
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dcc
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Dcc
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1,5-Diphenylcarbazide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
ProteoSpec™ N,N-Dicyclohexylcarbodiimide, Ricca Chemical
CAS: 538-75-0 Molecular Formula: C13H22N2 Molecular Weight (g/mol): 206.333 InChI Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N Synonym: dicyclohexylcarbodiimide,n,n'-dicyclohexylcarbodiimide,dccd,1,3-dicyclohexylcarbodiimide,dcci,carbodicyclohexylimide,bis cyclohexyl carbodiimide,dcc,n,n-dicyclohexylcarbodiimide,carbodiimide, dicyclohexyl PubChem CID: 10868 ChEBI: CHEBI:53090 IUPAC Name: N,N'-dicyclohexylmethanediimine SMILES: C1CCC(CC1)N=C=NC2CCCCC2