Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoSpec™ N,N-Dicyclohexylcarbodiimide, Ricca Chemical
SDP

Catalog No. MB135025C2 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MB135025C2 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB135025C2 Supplier Ricca Chemical Company Supplier No. RMB135025C

missing translation for 'validMainframeMsg'

Protein Crosslinker

Chemical Identifiers

CAS 538-75-0
Molecular Formula C13H22N2
Molecular Weight (g/mol) 206.333
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
PubChem CID 10868
ChEBI CHEBI:53090
IUPAC Name N,N'-dicyclohexylmethanediimine
SMILES C1CCC(CC1)N=C=NC2CCCCC2

Specifications

CAS Min % 100
CAS Max % 100
Melting Point 122°C
Color White
Boiling Point 34°C
Packaging Amber Glass Bottle
Quantity 5 g
Solubility Information Insoluble in water
Formula Weight 206.33
Percent Purity 99.0% min.
Grade Reagent
Physical Form Solid
Assay GC
Chemical Name or Material ProteoSpec™ N,N-dicyclohexylurea, DCC
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient Health
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.