accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHCl

CAS RN 5382-18-3

IUPAC Name: 4-chloropiperidine
Synonyms: 4-chloropiperidine
Molecular Weight (g/mol): 119.59
Molecular Formula: C5H10ClN
InChi Key: GBPBXBUHZSOKTH-UHFFFAOYSA-N
SMILES: ClC1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results