accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3CN

CAS RN 53833-30-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results