Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Ethyl-4,5-dimethyl-1,3-oxazole

Catalog No. CDS000714 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS000714 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS000714 Supplier Sigma Aldrich Supplier No. CDS000714100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-(ethylaminomethyl)pyridine is a secondary amine.4-(ethylaminomethyl)pyridine is suitable for use in the synthesis of sodium dithiocarbamate ligands
  • It may be used in the synthesis of 4-(ethylaminodithiocarbamate) methylpyridine diorganotin derivatives
  • It may be used in the synthesis of {[ethyl(pyridin-4-ylmethyl)carbamothioyl]sulfanido-α2 s,sα}(1,4,7,10,13,16-hexaoxacyclooctadecane-α6o)potassium

Specifications

CAS 53833-30-0
Molecular Formula C7H11NO
Linear Formula C7H11NO
MDL Number MFCD00036664
Quantity 10 g
Molecular Weight (g/mol) 125.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.