accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOH(Z)(Z)(Z)(Z)

CAS RN 53847-30-6

IUPAC Name: 1,3-dihydroxypropan-2-yl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Synonyms: 2-arachidonoylglycerol 2-ara-gl
Molecular Weight (g/mol): 378.55
Molecular Formula: C23H38O4
InChi Key: RCRCTBLIHCHWDZ-DOFZRALJSA-N
SMILES: CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)