accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(R)(S)OHNN(R)HONNH

CAS RN 53910-25-1

IUPAC Name: (8R)-3-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H,4H,7H,8H-imidazo[4,5-d][1,3]diazepin-8-ol
Synonyms: pentostatin 2-deoxycoformycin wyeth brand of pentostatin pentostatin (product) pentostatin preparation (R)-2'-deoxycoformycin prasfarma brand of pentostatin lederle brand of pentostatin 8R-2'-deoxycoformycin pentostatin (substance)
Molecular Weight (g/mol): 268.27
Molecular Formula: C11H16N4O4
InChi Key: FPVKHBSQESCIEP-JQCXWYLXSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1NC=NC[C@H]2O
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results