accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3H3CCH3

CAS RN 5396-38-3

IUPAC Name: 1-tert-butyl-4-methoxybenzene
Synonyms: 1-tert-butyl-4-methoxybenzene
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: MCUPBIBNSTXCPQ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results