accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5397-31-9

IUPAC Name: 3-[(2-ethylhexyl)oxy]propan-1-amine
Synonyms: 3-[(2-ethylhexyl)oxy]propan-1-amine
Molecular Weight (g/mol): 187.33
Molecular Formula: C11H25NO
InChi Key: DVFGEIYOLIFSRX-UHFFFAOYNA-N
SMILES: CCCCC(CC)COCCCN
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results