Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Ethylhexyloxy)propylamine 98.0+%, TCI America™
SDP

Catalog No. E0295500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0295500ML 500 mL
E029525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0295500ML Supplier TCI America Supplier No. E0295500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5397-31-9
Molecular Formula C11H25NO
Molecular Weight (g/mol) 187.33
MDL Number MFCD00025619
InChI Key DVFGEIYOLIFSRX-UHFFFAOYNA-N
Synonym 2-ethylhexyloxypropylamine, 3-2-ethylhexyloxy propylamine, 2-ethylhexyl 3-aminopropyl ether, 3-2-ethylhexyl oxy propan-1-amine, 3-2-ethylhexoxy propylamine, 3-2-ethylhexyl oxy propylamine, propylamine, 3-2-ethylhexoxy, 1-propanamine, 3-2-ethylhexyl oxy, 3-isooctoxyl propylamine
PubChem CID 21499
IUPAC Name 3-[(2-ethylhexyl)oxy]propan-1-amine
SMILES CCCCC(CC)COCCCN

Specifications

Color Colorless
Boiling Point 78°C
Quantity 500 mL
UN Number 2810
Formula Weight 187.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(2-Ethylhexyloxy)propylamine
EINECSNumber (2)-0385&(2)-0389
RTECSNumber UI2820000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.