accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSBrN

CAS RN 54045-74-8

IUPAC Name: methyl 2-bromo-1,3-thiazole-5-carboxylate
Synonyms: methyl 2-bromo-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 222.06
Molecular Formula: C5H4BrNO2S
InChi Key: HNLVIKMXFBRZDF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(Br)S1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)