accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONOO

CAS RN 5405-53-8

IUPAC Name: 2,7-dinitro-9H-fluorene
Synonyms: 9H-fluorene, 2,7-dinitro-
Molecular Weight (g/mol): 256.22
Molecular Formula: C13H8N2O4
InChi Key: IHZCVUBSTYOFSJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2C(CC3=CC(=CC=C23)[N+]([O-])=O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)