accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH2N

CAS RN 5407-87-4

IUPAC Name: 4,6-dimethylpyridin-2-amine
Synonyms: 2-pyridinamine, 4,6-dimethyl-
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: BRBUBVKGJRPRRD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(N)=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)