Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,6-dimethylpyridine, 99%

Catalog No. AC103580050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC103580050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC103580050 Supplier Thermo Scientific Chemicals Supplier No. 103580050

missing translation for 'validMainframeMsg'

CAS: 5407-87-4 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5407-87-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
InChI Key BRBUBVKGJRPRRD-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine
PubChem CID 21507
IUPAC Name 4,6-dimethylpyridin-2-amine
SMILES CC1=CC(=NC(=C1)N)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 65.0°C to 69.5°C
Color White to Brown
Boiling Point 235.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2(C5H2N)NH2
Quantity 5 g
Solubility Information Solubility in water: 5.5 g/100mL (20 c).
Formula Weight 122.17
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4, 6-dimethylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
EINECSNumber 226-470-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.