Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2,4-lutidine 98.0+%, TCI America™
SDP

Catalog No. A073125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A073125G 25 g
A07315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A073125G Supplier TCI America Supplier No. A073125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5407-87-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006322
InChI Key BRBUBVKGJRPRRD-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine
PubChem CID 21507
IUPAC Name 4,6-dimethylpyridin-2-amine
SMILES CC1=CC(=NC(=C1)N)C

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 231°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2,4-lutidine
RTECSNumber US1818000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.