accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 54105-66-7

IUPAC Name: undecylcyclohexane
Synonyms: undecylcyclohexane
Molecular Weight (g/mol): 238.46
Molecular Formula: C17H34
InChi Key: XQQVOBPJWHQXEN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results