accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNH2NHNN

CAS RN 5418-95-1

IUPAC Name: N''-(1H-1,3-benzodiazol-2-yl)guanidine
Synonyms: 2-benzimidazolylguanidine
Molecular Weight (g/mol): 175.2
Molecular Formula: C8H9N5
InChi Key: JJWCTKUQWXYIIU-UHFFFAOYSA-N
SMILES: NC(N)=NC1=NC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results