accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 542-75-6

IUPAC Name: (1Z)-1,3-dichloroprop-1-ene
Synonyms: telone II C3H4cl2 ccris 227 cis-DCP
Molecular Weight (g/mol): 110.97
Molecular Formula: C3H4Cl2
InChi Key: UOORRWUZONOOLO-UPHRSURJSA-N
SMILES: ClC\C=C/Cl
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)