Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-1-Propene (Mixed) Solution, 5000μg/mL in Methanol, Ultra Scientific

Catalog No. USEPA1034
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1034 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1034 Supplier Agilent Technologies Supplier No. EPA10341
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 542-75-6 , 67-56-1
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-UPHRSURJSA-N
PubChem CID 24726
ChEBI CHEBI:18624
IUPAC Name (1Z)-1,3-dichloroprop-1-ene
SMILES ClC\C=C/Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material 1, 3-Dichloro-1-Propene, 5000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.