Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloropropene, Mixture of Isomers 96%, Thermo Scientific™

Catalog No. AC210311000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC210311000 100 mL
AC210310250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210311000 Supplier Thermo Scientific Chemicals Supplier No. 210311000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-75-6
Molecular Formula C3H4Cl2
Molecular Weight (g/mol) 110.97
MDL Number MFCD00000985
InChI Key UOORRWUZONOOLO-UPHRSURJSA-N
Synonym 1,3-dichloropropene, trans-1,3-dichloropropene, telone ii, telone, 1,3-dichloropropylene, nemex, trans-1,3-dichloropropylene, dorlone ii, telone c, di-trapex cp
PubChem CID 24726
ChEBI CHEBI:18624
IUPAC Name (1Z)-1,3-dichloroprop-1-ene
SMILES ClC\C=C/Cl

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.2
Infrared Spectrum Authentic
Refractive Index 1.4705 to 1.4745 (20°C, 589nm)
Linear Formula ClCH2CH=CHCl
Quantity 100 mL
Beilstein 01, 199
Merck Index 15, 3085
Formula Weight 110.97
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloropropene, mixture of isomers, 96%
Show More Show Less
Hazard Category CANCER SUSPECTED AGENT
EINECSNumber 208-826-5
RTECSNumber UC8310000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.