accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3CNCH3OS

CAS RN 5424-47-5

IUPAC Name: hydrogen 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one chloride
Synonyms: hydrogen 3-(dimethylamino)-1-(thiophen-2-yl)propan-1-one chloride
Molecular Weight (g/mol): 219.73
Molecular Formula: C9H14ClNOS
InChi Key: HPVHJPMLORARSR-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCC(=O)C1=CC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results