accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3NH2

CAS RN 543-82-8

IUPAC Name: 6-methylheptan-2-amine
Synonyms: amidrine 2-metil-6-amino-eptano 2-isooctyl amine octodrine (inn) and its salts vaporpac α,epsilon-dimethylhexylamine octodrinum ottodrina 奥托君 octodrine
Molecular Weight (g/mol): 129.25
Molecular Formula: C8H19N
InChi Key: QNIVIMYXGGFTAK-UHFFFAOYNA-N
SMILES: CC(C)CCCC(C)N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results