accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOBr

CAS RN 54322-31-5

IUPAC Name: 4-(2-bromoethyl)benzene-1-sulfonic acid
Synonyms: 4-(2-bromoethyl)benzenesulfonic acid
Molecular Weight (g/mol): 265.12
Molecular Formula: C8H9BrO3S
InChi Key: XRPCDXSRXPQUKT-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=CC=C(CCBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results