accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3CH3

CAS RN 54396-97-3

IUPAC Name: 2-ethoxyethyl 2-methylpropanoate
Synonyms: 2-ethoxyethyl isobutyrate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: ANSOCBLHUNAGRK-UHFFFAOYSA-N
SMILES: CCOCCOC(=O)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results