CAS RN 544-31-0
IUPAC Name:
N-(2-hydroxyethyl)hexadecanamide
Synonyms:
palmitoylethanolamide palmitoyl-EA anandamide (16:0) palmitamide mea palmitoyl ethanolamide
Molecular Weight (g/mol):
299.5
Molecular Formula:
C18H37NO2
InChi Key:
HXYVTAGFYLMHSO-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC(=O)NCCO
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals Palmitoylethanolamide, 98%
Thermo Scientific Chemicals Palmitoylethanolamide, 98%
CAS: 544-31-0 Molecular Formula: C18H37NO2 Synonym: PEA; N-(2-Hydroxyethyl)hexadecanamide
2
R&D Systems™ Palmitoylethanolamide
CAS: 544-31-0 Molecular Formula: C18H37NO2