accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHOH

CAS RN 544-62-7

IUPAC Name: 3-(octadecyloxy)propane-1,2-diol
Synonyms: batyl alcohol batyl alcohol, (s)-isomer batilol batyl alcohol, (r)-isomer batyl alcohol, (dl)-isomer batyl alcohol, (DL)-isomer (s)-batyl alcohol batyl alcohol, (DL)- (R)-isomer batyl alcohol α-octadecylether of glycerol
Molecular Weight (g/mol): 344.58
Molecular Formula: C21H44O3
InChi Key: OGBUMNBNEWYMNJ-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCCOCC(O)CO
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results