Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Batyl Alcohol, MP Biomedicals

Catalog No. ICN213460 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN213460 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN213460 Supplier MP Biomedicals, Inc Supplier No. 0521346001

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 544-62-7
Molecular Formula C21H44O3
Molecular Weight (g/mol) 344.58
InChI Key OGBUMNBNEWYMNJ-UHFFFAOYSA-N
Synonym batyl alcohol, batilol, 1-o-octadecylglycerol, stearyl monoglyceride, batilolum, 3-octadecyloxy propane-1,2-diol, 3-octadecyloxy-1,2-propanediol, glycerine 1-monostearyl ether, monooctadecyl ether of glycerol, batylalcohol
PubChem CID 3681
ChEBI CHEBI:34117
IUPAC Name 3-octadecoxypropane-1,2-diol
SMILES CCCCCCCCCCCCCCCCCCOCC(CO)O

Specifications

Quantity 1 g
Formula Weight 344.58
Chemical Name or Material Batyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.