accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54454-12-5

IUPAC Name: 3-chloro-2-methylbenzonitrile
Synonyms: 3-chloro-2-methylbenzonitrile
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: FKFZTNLSUJCIMG-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Cl)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results