CAS RN 5449-69-4
IUPAC Name:
(2-methoxyphenyl)(4-methoxyphenyl)methanone
Synonyms:
(2-methoxyphenyl)(4-methoxyphenyl)methanone
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
QWWJLMQOKZOTNX-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(=O)C1=CC=CC=C1OC
1
–
1
of
1
results
1
2,4'-Dimethoxybenzophenone 98.0+%, TCI America™
CAS: 5449-69-4 Molecular Formula: C15H14O3 Molecular Weight (g/mol): 242.274 MDL Number: MFCD01311508 InChI Key: QWWJLMQOKZOTNX-UHFFFAOYSA-N PubChem CID: 226205 IUPAC Name: (2-methoxyphenyl)-(4-methoxyphenyl)methanone SMILES: COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2OC