accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 545-06-2

IUPAC Name: trichloroacetonitrile
Synonyms: trichloroacetonitrile
Molecular Weight (g/mol): 144.38
Molecular Formula: C2Cl3N
InChi Key: DRUIESSIVFYOMK-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C#N
Molecular Weight (g/mol) 
  • (8)
  • (4)
  • (3)
Boiling Point 
  • (3)
  • (4)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (1)
  • (3)
  • (7)
  • (8)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (7)
  • (7)
  • (1)