Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetonitrile Solution, 1000μg/mL in Acetone, Ultra Scientific

Catalog No. USEPA1239A
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USEPA1239A 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USEPA1239A Supplier Agilent Technologies Supplier No. EPA1239A1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 545-06-2 , 67-64-1
Molecular Formula C2Cl3N
Molecular Weight (g/mol) 144.379
InChI Key DRUIESSIVFYOMK-UHFFFAOYSA-N
PubChem CID 11011
ChEBI CHEBI:82541
IUPAC Name 2,2,2-trichloroacetonitrile
SMILES C(#N)C(Cl)(Cl)Cl

Specifications

CAS Max % 99.87, 0.13
Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Trichloroacetonitrile Solution, 1000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.