accessibility menu, dialog, popup

UserName

Viewing profile as user:
OI

CAS RN 54555-84-9

IUPAC Name: [(2-iodoethoxy)methyl]benzene
Synonyms: [(2-iodoethoxy)methyl]benzene
Molecular Weight (g/mol): 262.09
Molecular Formula: C9H11IO
InChi Key: NYACRWGQRUIHSO-UHFFFAOYSA-N
SMILES: ICCOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results