accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 54567-92-9

IUPAC Name: 2,2-dichloro-1,1,1-triethoxyethane
Synonyms: 2,2-dichloro-1,1,1-triethoxyethane
Molecular Weight (g/mol): 231.11
Molecular Formula: C8H16Cl2O3
InChi Key: ZKMPARWUDMKPMZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)(OCC)C(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results