Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Orthodichloroacetate, TCI America™
SDP

Catalog No. O02741G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02741G Supplier TCI America Supplier No. O02741G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 54567-92-9
Molecular Formula C8H16Cl2O3
Molecular Weight (g/mol) 231.113
MDL Number MFCD01861293
InChI Key ZKMPARWUDMKPMZ-UHFFFAOYSA-N
Synonym 1,1,1-Triethoxy-2,2-dichloroethane, Orthodichloroacetic Acid Triethyl Ester
PubChem CID 12723675
IUPAC Name 2,2-dichloro-1,1,1-triethoxyethane
SMILES CCOC(C(Cl)Cl)(OCC)OCC

Specifications

Color Colorless
Boiling Point 195°C
Quantity 1 g
Formula Weight 231.11
Physical Form Liquid
Chemical Name or Material Triethyl Orthodichloroacetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.