accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5458-59-3

IUPAC Name: propan-2-yl octanoate
Synonyms: isopropyl octanoate
Molecular Weight (g/mol): 186.3
Molecular Formula: C11H22O2
InChi Key: WCGIIHOFOFCKSM-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results