accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 5465-86-1

IUPAC Name: 4-methyl-2-(piperidin-1-yl)quinoline
Synonyms: 4-methyl-2-(piperidin-1-yl)quinoline
Molecular Weight (g/mol): 226.32
Molecular Formula: C15H18N2
InChi Key: USYRQXDHKXGTCK-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=NC(=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)