accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNSSH2ONaNa(E/Z)

CAS RN 5466-54-6

IUPAC Name: disodium (1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide hydrate
Synonyms: disodium (1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide hydrate
Molecular Weight (g/mol): 204.17
Molecular Formula: C4H2N2Na2OS2
InChi Key: DWYJCSXARKJSBC-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].[S-]C(C#N)=C([S-])C#N
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)