accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 547-32-0

IUPAC Name: sodium 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
Synonyms: sulfadiazine sodium
Molecular Weight (g/mol): 272.26
Molecular Formula: C10H9N4NaO2S
InChi Key: JLDCNMJPBBKAHH-UHFFFAOYSA-N
SMILES: [Na+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1
Percent Purity 
  • (2)
  • (6)
Color 
  • (6)
Grade 
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)