CAS RN 547-57-9
Thermo Scientific Chemicals Tropaeolin O
CAS: 547-57-9 Molecular Formula: C12H10N2NaO5S Molecular Weight (g/mol): 317.271 MDL Number: MFCD00007499 InChI Key: YOGUPPQYRWYKGT-IRIIKGHASA-N Synonym: Acid Orange 6; C.I. 14270 PubChem CID: 87071197 IUPAC Name: 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium SMILES: C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]
Thermo Scientific Chemicals Tropaeolin O, 0.04% w/v aq. soln.
CAS: 547-57-9 Molecular Formula: C12H10N2NaO5S Molecular Weight (g/mol): 317.271 MDL Number: MFCD00007499 InChI Key: YOGUPPQYRWYKGT-IRIIKGHASA-N Synonym: tropaeolin o PubChem CID: 87071197 IUPAC Name: 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium SMILES: C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]
Thermo Scientific Chemicals Tropaeolin O, pure
CAS: 547-57-9 Molecular Formula: C12H10N2NaO5S Molecular Weight (g/mol): 317.271 MDL Number: MFCD00007499 InChI Key: YOGUPPQYRWYKGT-IRIIKGHASA-N Synonym: Acid orange 6,C.I. 14270,4-(2, 4-Dihydroxyphenylazo)benzenesulfonic acid, sodium salt PubChem CID: 87071197 IUPAC Name: 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium SMILES: C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]
Sulfo Orange Indicator, 0.04% (w/v), Ricca Chemical
CAS: 547-57-9
Tropaeolin O, TCI America™
CAS: 547-57-9 Molecular Formula: C12H10N2NaO5S Molecular Weight (g/mol): 317.271 MDL Number: MFCD00007499 InChI Key: YOGUPPQYRWYKGT-IRIIKGHASA-N Synonym: tropaeolin o PubChem CID: 87071197 IUPAC Name: 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium SMILES: C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]
Tropaeolin O Sodium Salt, Solstice
CAS: 547-57-9 Molecular Formula: C12H10N2NaO5S Molecular Weight (g/mol): 317.271 MDL Number: MFCD00007499 InChI Key: YOGUPPQYRWYKGT-IRIIKGHASA-N PubChem CID: 87071197 IUPAC Name: 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium SMILES: C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]