Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tropaeolin O, TCI America™
SDP

Catalog No. R00101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00101G Supplier TCI America Supplier No. R00101G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 547-57-9
Molecular Formula C12H10N2NaO5S
Molecular Weight (g/mol) 317.271
MDL Number MFCD00007499
InChI Key YOGUPPQYRWYKGT-IRIIKGHASA-N
Synonym tropaeolin o
PubChem CID 87071197
IUPAC Name 4-[(2Z)-2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzenesulfonic acid;sodium
SMILES C1=CC(=CC=C1NN=C2C=CC(=O)C=C2O)S(=O)(=O)O.[Na]

Specifications

Quantity 1 g
Formula Weight 316.26
Physical Form Crystalline Powder
Chemical Name or Material Tropaeolin O
EINECSNumber (5)-1454
RTECSNumber DB6145180
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.