accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFSOOOFFFSOOOFFFSOOOYb

CAS RN 54761-04-5

IUPAC Name: ytterbium(3+) tritrifluoromethanesulfonate
Synonyms: ytterbium(3+) tritriflate
Molecular Weight (g/mol): 620.24
Molecular Formula: C3F9O9S3Yb
InChi Key: AHZJKOKFZJYCLG-UHFFFAOYSA-K
SMILES: [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results