CAS RN 548-19-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ Pre-mRNA Splicing Inhibitor, Isoginkgetin
The Pre-mRNA Splicing Inhibitor, Isoginkgetin, also referenced under CAS 548-19-6, blocks the spliceosome-meidated splicing process.
2
Tocris Bioscience™ Isoginkgetin
Pre-mRNA splicing inhibitor; cell permeable